missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 2mm Thick

For use in preparatory electrophoresis and high-throughput screening.

Brand:  Fisherbrand™ VS10-16MC-2

 View more versions of this product

Product Code. 11827603


Description

Description

  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Recommended for maximising sample volume
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications

Specifications

Blue
OmniPAGE Mini Vertical Gel Electrophoresis Unit
16
2 mm
Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations
Comb
1 Each
50 μL
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Fisherbrand and Fisher BioReagents Focus on electrophoresis

    SDS
    Documents

    Documents

    Certificates
    Promotions

    Promotions

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 2mm Thick >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.