Learn More
WTW™ Cleaning Solution for WTW™ Galvanic Oxygen Sensors StirrOx G™, CellOx 325™, DurOx 325™
Get optimum results with this cleaning solution that allows for self-stirring dissolved oxygen sensor StirrOx™ G for the measurement of the biochemical oxygen demand (BOD) according to the dilution method.
Brand: WTW™ 205204
73.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Cleaning solution for galvanic oxygen sensors
- Cleaning solution for StirrOx™ self-stirring dissolved oxygen sensor
- Cleaning solution that allows for self-stirring dissolved oxygen sensor StirrOx™ G for the measurement of the biochemical oxygen demand (BOD) according to the dilution method
- For use with cleaning galvanic oxygen sensors StirrOx™ G, CellOx™325, DurOx™ 325, TA 197 Oxi, ConOx, MPP 350 D.O. module

Specifications
Clear | |
Cleaning solution |
Safety and Handling
- RL-G
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : acetic acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.