Learn More
Masterflex™ Chem-Durance Bio L/S™ Two-Stop Pump Tubing
Excellent chemical resistance and long pumping life
Brand:  Masterflex™ 96117-16
 View more versions of this product
Product Code. 16438743
  • 217.00€ / Pack of 8
Estimated Shipment: 10-10-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Meets USP Class VI requirements
  • Flexible outer jacket acts as an insulating layer to absorb and dissipate heat from the rollers - extending the life of the tubing
  • Plasticizer-free, smooth bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids
Specifications
Specifications
3.1 mm
1.25 ft.
Laboratory, Pharmaceutical Processing, and Biotech
8
460 mL/min
40 psi, 2.7 bar
40°C
Chem-Durance Bio
0.38 m
0°C
16
1/8 in.
660 mm Hg
2-stop Tubing
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Masterflex™ Chem-Durance Bio L/S™ Two-Stop Pump Tubing > 16