Learn More
Fisherbrand™ Powder Funnels
Product Code. 11907914 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric)Top:
6.5cm
7.5cm
10.5cm
15cm
Diameter (Metric) Stem:
16mm
21mm
27mm
Capacity (Metric):
50 mL
100 mL
225 mL
750 mL
Height (Metric):
6.6 cm
8.4 cm
10.3 cm
13.7 cm
Unit Size:
Pack of 24
Pack of 36
Pack of 4
Pack of 6
Product Code. 11907914
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Parallel stems minimize bridging of powder.
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 15cm
Capacity (Metric) 750 mL
Diameter (Metric) Stem 27mm
Material Polypropylene
Height (Metric) 13.7 cm
Autoclavable Autoclavable
Length (Metric) Stem 30mm