Learn More
IBA Lifesciences CD8 Fab-TACS™ Agarose Column Starter Kit
Label-free isolation of CD8+ T cells
154.00€ - 265.50€
Specifications
Buffer 10 x Buffer Cl
View More Specs
Products 2
Product Code Brand Target Species Donor Source Includes Price Quantity & Availability  
Product Code Brand Target Species Donor Source Includes Price Quantity & Availability  
17237740
View Documents
IBA Lifesciences
6-3203-002
Human Whole blood, buffy coat or PBMCs Strep-Tactin™ TACS Agarose columns, 1 mL (2 x), CD8 Fab-Strep (human), 10 x Buffer CI, Biotin stock solution, TACS Column Adapter This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
17287740
View Documents
IBA Lifesciences
6-3305-002
Mouse Murine splenocytes Strep-Tactin™ TACS Agarose columns, 0.3 mL (2 x), CD8 Fab-Strep (mouse), 10 x Buffer CI, Biotin stock solution, TACS Column Adapter This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description
Description
  • Fab-TACS™ - Traceless Affinity Cell Selection - is a positive cell isolation approach based on our Strep-tag™ technology.
  • Strep-Tactin™ TACS Agarose columns are filled with Strep-Tactin™ coated agarose macrobeads developed for cell isolation. Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) bind to the matrix.
  • Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to a surface marker. Non-target cells are efficiently washed away.
  • In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps.
  • After elution, the Fab-Streps self-dissociate from the cell surfaces due to their reduced affinity.
  • The label-free authentic target cells are now ready for further downstream applications
Specifications
Specifications
10 x Buffer Cl
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only.