Learn More
IBA Lifesciences CD8 Fab-TACS™ Agarose Column Starter Kit
Label-free isolation of CD8+ T cells
Brand:  IBA Lifesciences 6-3305-002
 View more versions of this product
Product Code. 17287740
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Fab-TACS™ - Traceless Affinity Cell Selection - is a positive cell isolation approach based on our Strep-tag™ technology.
  • Strep-Tactin™ TACS Agarose columns are filled with Strep-Tactin™ coated agarose macrobeads developed for cell isolation. Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) bind to the matrix.
  • Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to a surface marker. Non-target cells are efficiently washed away.
  • In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps.
  • After elution, the Fab-Streps self-dissociate from the cell surfaces due to their reduced affinity.
  • The label-free authentic target cells are now ready for further downstream applications
Specifications
Specifications
Strep-Tactin™ TACS Agarose columns, 0.3 mL (2 x), CD8 Fab-Strep (mouse), 10 x Buffer CI, Biotin stock solution, TACS Column Adapter
Murine splenocytes
Mouse
10 x Buffer Cl
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only.