Learn More
CD31 Rat anti-Mouse, BUV615, Clone: 390, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 751047
657.57 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
CD31 Monoclonal antibody specifically detects CD31 in Mouse samples. It is validated for Flow Cytometry
Specifications
CD31 | |
Monoclonal | |
Brilliant Ultraviolet 615 | |
Platelet endothelial cell adhesion molecule; Pecam1; PECAM-1; Pecam-1 | |
C3H/HeJ mouse hematopoietic progenitor cell line 32D | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
390 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.