Learn More
Greiner Bio-One ThinCert™ Tissue Culture Inserts
Research tool for transport, co-culture, migration and invasion studies. Greiner Bio-One ThinCert™ Tissue Culture Inserts provides improved cell adhesion due to physical surface treatment.
Brand: Greiner Bio-One 657630
This item is not returnable.
View return policy
Description
- Clear polystyrene housing with sealed PET capillary pore membrane
- Hanging geometry provides protection to lower compartment cell layer
- Simplified pipetting due to self-lift geometry
- Eccentric in-well positioning allows unequalled access to lower compartment
- Available in 6, 12 and 24 well format
Specifications
Polystyrene housing with sealed PET capillary pore membrane | |
0.6 x 106 pores/cm2 | |
3 μm | |
6 |
Product Content Correction
Product Title
Greiner Bio-One ThinCert™ Tissue Culture Inserts > Format: 6 well; Pore size: 3μm; Pore Density: 0.6 x 106 cm2