missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Calcium Ionophore

Calcium Ionophore is an antibiotic that possesses weak in vitro activity against gram positive organisms.

Brand:  Novus Biologicals™ NBP2-33206-1MG

Product Code. 18117012

  • 138.00€ / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Application Notes: Ionophore is highly selective for Ca2+. Potentiates responses to NMDA but not quisqualate. In cell culture, stimulates nitric oxide production by calmodulin-dependent constitutive nitric oxide synthase.

Solubility: Ethanol: 5 mg/mL (with heating. Solutions are stable for at least 3 months at -20°C when protected from light and water contamination.) DMSO: 50 mg/mL (with heating or sonication. Solutions are stable for at least 3 months at -20°C when protected from light and water contamination.) Aqueous solutions can be made by diluting DMSO solutions in H2O.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 4°C in the dark.
1 mg
Calcium Ionophore
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Calcium Ionophore > 1 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.