missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetanus Toxin Antibody, DyLight 550, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-63239R
This item is not returnable.
View return policy
Description
Tetanus Toxin Polyclonal antibody specifically detects Tetanus Toxin in Bacteria samples. It is validated for Western Blot, ELISA
Specifications
Tetanus Toxin | |
Polyclonal | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 550 | |
Tentoxylysin | |
Purified tetanus toxin | |
0.1 mL | |
Virology Bacteria and Parasites | |
Bacteria | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tetanus Toxin Antibody, DyLight 550, Novus Biologicals™ > 0.1 mL; DyLight 550
Spot an opportunity for improvement?Share a Content Correction