Learn More
Trajan Scientific and Medical™ BP1 GC Capillary Columns
Suitable for all routine analysis. Trajan Scientific and Medical BP1 GC Capillary Columns are industry standard non polar phase columns.
Brand: Trajan Scientific and Medical™ 054043
This item is not returnable.
View return policy
Description
- Classic cross-linked dimethyl polysiloxane technology
- Low bleed
- Non-polar
- Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
0.25 mm | |
0.25 mm | |
-60°C to +320/340°C | |
15 m | |
BP1 |
BP1 GC Column | |
Amines Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines | |
0.25 μm | |
100% Dimethyl Polysiloxane |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Trajan Scientific and Medical™ BP1 GC Capillary Columns > Length: 15m; Dia.: 0.25mm; Thickness: 0.25μm