Learn More
Trajan Scientific and Medical™ BP1 GC Capillary Columns
Suitable for all routine analysis. Trajan Scientific and Medical BP1 GC Capillary Columns are industry standard non polar phase columns.
Brand: Trajan Scientific and Medical™ 054810
This item is not returnable.
View return policy
Description
- Classic cross-linked dimethyl polysiloxane technology
- Low bleed
- Non-polar
- Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
0.32 mm | |
Hydrocarbon Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines | |
1 μm | |
100% Dimethyl Polysiloxane |
BP1 GC Column | |
-60°C to +340/360°C | |
60 m | |
BP1 |
Product Content Correction
Product Title
Trajan Scientific and Medical™ BP1 GC Capillary Columns > Length: 60m; Dia.: 0.32mm; Thickness: 1μm