Learn More
Rogo Sampaic™ Class B Glass Burettes
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Rogo Sampaic™ Class B Glass Burettes are manufactured from durable clear borosilicate glass.
Brand: Rogo Sampaic™ DND010
This item is not returnable.
View return policy
Description
- With funnel
- Straight glass faucet
- Permanent red graduation marks
- Corrosion-free and resistant to thermal shock
Specifications
50 mL | |
Borosilicate Glass | |
0.10 mL | |
0.10 mL |
Product Content Correction
Product Title
Rogo Sampaic™ Class B Glass Burettes > Capacity: 50 mL