missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOK Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41045MFV610
This item is not returnable.
View return policy
Description
BOK Polyclonal antibody specifically detects BOK in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
BOK | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Bcl2-L-9, BCL2L9bcl-2-related ovarian killer protein, Bcl-2-like protein 9, BCL2-related ovarian killer, BOKL, hBOK, MGC4631 | |
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human BOK. The immunogen is located within the first 50 amino acids of BOK. Amino Acid Squence: DAFDRSPTDKELVAQA | |
0.1 mL | |
Apoptosis | |
666 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50 mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BOK Antibody, mFluor Violet 610 SE, Novus Biologicals™ > 0.1 mL, mFluor Violet 610 SE
Spot an opportunity for improvement?Share a Content Correction