Learn More
Genevac™ miVac Centrifugal Concentrators
Choose the ideal concentrator according to workload. Genevac™ miVac Centrifugal Concentrators are modular systems for removing water and solvents from samples. They are available with a range of unique JetRotors™ and vacuum pumps. An optional high-power cold trap is used to condense solvent vapors.
Brand:  Genevac™ DNA-23050-B00
 View more versions of this product
Product Code. 11574604
  • 12215.00€ / Each
Explore more special offers
Récupel-bijdrage uitgesloten
Description
Show More
Specifications
Specifications
miVac DNA
Samples in tubes, microplates and vials
Concentrator with concentration chamber, low-vacuum pump, tube rotor
360W x 597D x 300H mm
230 V 50/60 Hz 650 w
30°C to 80°C
120 Reaction Vessels (0.5 mL), 6 Microtiter Plates
Max G-force: 250
34 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Genevac™ miVac Centrifugal Concentrators > Model: miVac DNA; Capacity: 120 reaction vessels 0.5mL or 6 microtiter plates; Includes concentrator with small chamber, pump, rotor