Learn More
BD Safety-Lok™ Blood Collection Set
Use for blood collection. BD Vacutainer™ Safety-Lok™ Blood Collection Set (50 sets) comes with safety re-shielding cover and Luer adapter.
Brand: BD 367282
Description
- Includes safety re-shielding cover and Luer adapter,
- 21 G ¾ in., 18 cm tubing, green
- Recommended for adults
- One case consists fo 4 x 50 Blood Collection Sets
- One Set is necessary for a pair of blood culture bottles
Specifications
Blood Collection Set | |
Sterile |