missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse BLC/CXCL13 ELISA Kit

ELISA

664.00€ - 2780.00€

Specifications

Accession Number O55038
Assay Range 1.37 to 1000 pg/mL
Assay Sensitivity 2 pg/mL
Calibration Range 2 pg/mL to 2000 pg/mL
Description BLC (CXCL13) ELISA Kit, 96 Assays, 1 Kit, Mouse Species, HRP Conjugate, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of BLC (CXCL13) in Samples Using a Microplate Reader
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15455253
View Documents
Invitrogen™
EMCXCL13
96 Tests
664.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
17175709
View Documents
Invitrogen™
EMCXCL13X5
5 x 96 Tests
2780.00€
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Mouse BLC/CXCL13 ELISA quantitates Ms BLC in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms BLC. Principle of the method The Mouse BLC solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

CXCL13, also known as BCA-1 and Angie, is a small chemokine strongly expressed in the secondary lymphoid organs. It preferentially promotes the migration of B lymphocytes (both B-1 and B-2) to the follicles of the spleen, lymph nodes, and Peyer's patches. It is produced by macrophages, and in mouse the highest constitutive producers include large peritoneal macrophages, Kuppfer cells and lung macrophages (both CD11c+ and CD11c-). The cellular ligand for CXCL13 is CXCR5 also known as BLR-1 (Burkitt’s Lymphoma Receptor 1). CXCR5 is expressed on B cells, Tfh cells and, to a lower degree, some memory T cell populations. CXCL13 plays an important role in the development of chronic lymphocytic leukemia (CCL). CXCL13 is also a plasma biomarker for germinal center activity and generation of broadly neutralizing antibodies against HIV.
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Mouse BLC/CXCL13 ELISA Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.