missing translation for 'onlineSavingsMsg'
Learn More
Beta - Nicotinamide Adenine Dinucleotide Phosphate Reduced Tetrasodium Salt 98% MP Biomedicals Product Code.: 11358779

Beta - Nicotinamide Adenine Dinucleotide Phosphate Reduced Tetrasodium Salt 98% MP Biomedicals

Product Code. 11358779 Shop All MP Biomedicals Products
100 mg
Click to view available options
Quantity:
25 mg
50 mg
100 mg
250 mg
500 mg
1 g
This item is not returnable. View return policy

Product Code. 11358779

Brand: MP Biomedicals 0210116780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2646-71-1
Molecular Formula C21H26N7Na4O17P3
Molecular Weight (g/mol) 833.35
MDL Number MFCD10567216
InChI Key WYWWVJHQDVCHKF-NRTBITFTNA-J
Synonym nadph tetrasodium salt, nadph, tetrasodium salt, coenzyme ii tetrasodium salt reduced form
PubChem CID 131673986

Specifications

Assay Percent Range 98%
Quantity 100 mg
SMILES NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP(O)(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
IUPAC Name [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
Formula Weight 833.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Beta - Nicotinamide Adenine Dinucleotide Phosphate, Reduced Tetrasodium Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Beta - Nicotinamide Adenine Dinucleotide Phosphate Reduced Tetrasodium Salt 98% MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.