missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta - Nicotinamide Adenine Dinucleotide Phosphate Reduced Tetrasodium Salt 98% MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
50 mg
100 mg
250 mg
500 mg
1 g
Specifications
Specifications
Color | White |
Assay Percent Range | 98% |
Quantity | 500 mg |
SMILES | [Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
IUPAC Name | [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium |
Formula Weight | 833.4 |
Percent Purity | 0.98 |
Physical Form | Powder |
Chemical Name or Material | β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, Reduced Tetrasodium Salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Beta - Nicotinamide Adenine Dinucleotide Phosphate Reduced Tetrasodium Salt 98% MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction