Learn More
behr Labor-Technik™ behrotest™ Extraction Device for Twisselmann Hot Extraction System
Extractors for multi-samples.
Brand: behr Labor-Technik™ B00218424
This item is not returnable.
View return policy
Description
- Extraction according to Twisselmann method
- Continuous hot extraction
- Improves solubility and shortens extraction time
- Energy individually adjustable for every sample position
- Cooling water distribution strip ensures uniform cooling at sample positions
- Practical condenser ledge to safely deposit condenser between hot extractions
- Brackets for safely holding intermediate extraction pieces after removing sample container
Specifications
Twisselmann Hot Extraction System | |
6 x 100 mL | |
Extraction of Solvents |
Extraction Device | |
Brackets |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
BEH011 behr Labor Technik Product Program 05 2008 Franzosisch
File size: 3.8 MB
BEH002 BEHR EQUIPEMENT FOR FOODSTUFFS
File size: 32.3 kB
BEH021 EXTRACTEUR SOXHLET 30 ET 100 ML
File size: 364.3 kB
Product Content Correction
Product Title
behr Labor-Technik™ behrotest™ Extraction Device for Twisselmann Hot Extraction System > Complete for six positions; Holds100mL round-bottomed flasks