Learn More
behr Labor-Technik™ behrotest™ Extraction Device for Twisselmann Hot Extraction System
Product Code. p-7083524
Click to view available options
Capacity (Metric):
6 x 100 mL
Unit Size:
Each
Récupel-bijdrage uitgesloten
Product Code. 10245152
Brand: behr Labor-Technik™ B00218424
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Extractors for multi-samples.
  • Extraction according to Twisselmann method
  • Continuous hot extraction
  • Improves solubility and shortens extraction time
  • Energy individually adjustable for every sample position
  • Cooling water distribution strip ensures uniform cooling at sample positions
  • Practical condenser ledge to safely deposit condenser between hot extractions
  • Brackets for safely holding intermediate extraction pieces after removing sample container
Specifications
For Use With (Equipment) Twisselmann Hot Extraction System
Product Type Extraction Device
Capacity (Metric) 6 x 100 mL
Includes Brackets
For Use With (Application) Extraction of Solvents