Learn More
Lenz Laborglasinstrumente™ Allihn Condensers with Glass Olives
Made of borosilicate glass. Lenz Laborglasinstrumente™ Allihn Condensers with Glass Olives are for condensation of vapors, with NS core and NS sleeve.
Brand:  Lenz Laborglasinstrumente™ 05223304
Product Code. 10313191
  • 88.00€ / Each
Estimated Shipment: 03-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • NS core and NS sleeve
  • Water connection
  • Made of DURAN™ pipe
Specifications
Specifications
Borosilicate Glass
250 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Allihn Condensers with Glass Olives > Length (Metric): 250mm