Learn More
Lenz Laborglasinstrumente™ Allihn Condensers with Glass Olives
Made of borosilicate glass. Lenz Laborglasinstrumente™ Allihn Condensers with Glass Olives are for condensation of vapors, with NS core and NS sleeve.
Brand: Lenz Laborglasinstrumente™ 05223304
This item is not returnable.
View return policy
Description
- NS core and NS sleeve
- Water connection
- Made of DURAN™ pipe
Specifications
Borosilicate Glass | |
250 mm |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Allihn Condensers with Glass Olives > Length (Metric): 250mm