Learn More
Witeg™ Allihn Bulb Condensers
Use the Witeg™ Allihn Bulb Condenser, which consists of a long glass tube with a water jacket. A series of bulbs on the tube increases the surface area.
Brand: Witeg™ 1 724 400
This item is not returnable.
View return policy
Description
- Various jacket lengths
Specifications
6.4 mm | |
400 mm | |
29/32 |
11 mm | |
1 Pack | |
Allihn Bulb Condenser |
Product Content Correction
Product Title
Witeg™ Allihn Bulb Condensers > Cone ST: 29/32; Length: 400mm