Learn More
Witeg™ Allihn Bulb Condensers
Use the Witeg™ Allihn Bulb Condenser, which consists of a long glass tube with a water jacket. A series of bulbs on the tube increases the surface area.
Brand:  Witeg™ 1 724 250
 View more versions of this product
Product Code. 11863562
  • 166.00€ / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Description
  • Various jacket lengths
Specifications
Specifications
6.4 mm
250 mm
29/32
11 mm
1 Pack
Allihn Bulb Condenser
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Witeg™ Allihn Bulb Condensers > Cone ST: 29/32; Length: 250mm