Learn More
BINDER™ Access Port
Access port with silicone plug for use with KB65, KB130, KB260, KB470, KB720, and KB 1060.
Brand: BINDER™ 8012-2467
This item is not returnable.
View return policy
Description
- Round access port with silicone plug
- Choose between 30, 50, or 100 mm plug
- Available on left, right, or top of the chamber
- For use with KB65, KB130, KB260, KB470, KB720, and KB 1060
Specifications
KB 65, KB 130, KB 260, KB 470, KB 720, KB 1060 | |
Access Port | |
100 mm |
Silicone | |
1 on Left Side |
Product Content Correction
Product Title
BINDER™ Access Port > Access Port, 100 mm, Left