missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Phosphate Cell Test
Brand: WTW™ 252021
144.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
EC No. 1907/2006 | |
Phosphate Cell Test | |
Reagent for analysis | |
25 Pack | |
The product is chemically stable under standard ambient conditions |
15°C to 25°C | |
Odorless |
|
Neutral |
|
9 to 307mg/L |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Phosphate Cell Test > Measuring Range: 9 to 307mg/L
Spot an opportunity for improvement?Share a Content Correction