missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 7-Chlorokynurenic acid sodium salt

Potent competitive inhibitor of L-glutamate uptake. Sodium salt of 7-Chlorokynurenic acid (Cat. No. 0237)

159.00€ - 555.00€

Specifications

Purity >98%
Product Type 7-Chlorokynurenic acid sodium salt
Molecular Weight (g/mol) 245.59
Inhibitors 7-Chlorokynurenic acid sodium salt
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18025892
View Documents
Tocris Bioscience™
3697/50
50 mg
555.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
18084182
View Documents
Tocris Bioscience™
3697/10
10 mg
159.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium salt of 7-Chlorokynurenic acid (Cat.No. 0237), an NMDA receptor antagonist acting at the glycine site. Potent competitive inhibitor of L-glutamate transport into synaptic vesicles.
Specifications

Specifications

>98%
7-Chlorokynurenic acid sodium salt
245.59
7-Chlorokynurenic acid sodium salt
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.