Learn More
Tocris Bioscience™ 7-Chlorokynurenic acid sodium salt
Potent competitive inhibitor of L-glutamate uptake. Sodium salt of 7-Chlorokynurenic acid (Cat. No. 0237)
Brand:  Tocris Bioscience™ 3697/10
 View more versions of this product
Product Code. 18084182
  • 159.00€ / 10mg
Explore more special offers
Description
Description
7-Chlorokynurenic acid sodium salt is a sodium salt of 7-Chlorokynurenic acid (Cat.No. 0237), an NMDA receptor antagonist acting at the glycine site. Potent competitive inhibitor of L-glutamate transport into synaptic vesicles.
Specifications
Specifications
>98%
7-Chlorokynurenic acid sodium salt
7-Chlorokynurenic acid sodium salt
10 mg
245.59