Learn More
Tocris Bioscience™ 7-Chlorokynurenic acid sodium salt
Potent competitive inhibitor of L-glutamate uptake. Sodium salt of 7-Chlorokynurenic acid (Cat. No. 0237)
Brand: Tocris Bioscience™ 3697/50
This item is not returnable.
View return policy
Description
7-Chlorokynurenic acid sodium salt is a sodium salt of 7-Chlorokynurenic acid (Cat.No. 0237), an NMDA receptor antagonist acting at the glycine site. Potent competitive inhibitor of L-glutamate transport into synaptic vesicles.Specifications
>98% | |
7-Chlorokynurenic acid sodium salt | |
7-Chlorokynurenic acid sodium salt |
50 mg | |
245.59 |