missing translation for 'onlineSavingsMsg'
Learn More
(4R)-(+)-4-Isopropyl-2-oxazolidinone, 98%, Thermo Scientific Chemicals Product Code.: 10470252

(4R)-(+)-4-Isopropyl-2-oxazolidinone, 98%, Thermo Scientific Chemicals

Product Code. 10470252
1 g, Glass Bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10470252

Brand: Thermo Scientific Acros 297850010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95530-58-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00075172
InChI Key YBUPWRYTXGAWJX-YFKPBYRVSA-N
Synonym r-4-isopropyl-2-oxazolidinone, r-4-isopropyloxazolidin-2-one, r-+-4-isopropyl-2-oxazolidinone, 4r-+-4-isopropyl-2-oxazolidinone, 4r-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4r, 4r-4-propan-2-yl-1,3-oxazolidin-2-one, pubchem8176, r-isopropyloxazolidinone
PubChem CID 641505
IUPAC Name (4R)-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)[C@@H]1COC(=O)N1

Specifications

Melting Point 70°C to 73°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Specific Rotation -18
Solubility Information Solubility in water: soluble in water
Specific Rotation Condition −18° (20°C c=1,ethanol)
Quantity 1 g
Formula Weight 129.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (4R)-(+)-4-Isopropyl-2-oxazolidinone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0374350 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number297850050,297850250,297850010
Lot NumberA0305690 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog Number297850050,297850250,297850010
Product Identifier
  • (4R)-(+)-4-Isopropyl-2-oxazolidinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(4R)-(+)-4-Isopropyl-2-oxazolidinone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.