missing translation for 'onlineSavingsMsg'
Learn More

(4R)-(+)-4-Isopropyl-2-oxazolidinone, 98+%, Thermo Scientific Chemicals

Product Code. 11313566
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11313566

Brand: Thermo Scientific Alfa Aesar L14518.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95530-58-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00075172
InChI Key YBUPWRYTXGAWJX-YFKPBYRVSA-N
Synonym r-4-isopropyl-2-oxazolidinone, r-4-isopropyloxazolidin-2-one, r-+-4-isopropyl-2-oxazolidinone, 4r-+-4-isopropyl-2-oxazolidinone, 4r-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4r, 4r-4-propan-2-yl-1,3-oxazolidin-2-one, pubchem8176, r-isopropyloxazolidinone
PubChem CID 641505
IUPAC Name (4R)-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)[C@@H]1COC(=O)N1

Specifications

Melting Point 70°C to 74°C
Beilstein 5376658
Quantity 1 g
Formula Weight 129.16
Optical Rotation +17° (c=6 in Ethanol)
Percent Purity ≥98%
Chemical Name or Material (4R)-(+)-4-Isopropyl-2-oxazolidinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.