missing translation for 'onlineSavingsMsg'
Learn More
2-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals Product Code.: 11487347

2-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11487347
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11487347

Brand: Thermo Scientific Alfa Aesar A15066.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

Solubility
Soluble in water(1.4g/L).

Notes
Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-30-9
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.993
MDL Number MFCD00001071
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0eShow MoreShow Less
PubChem CID 6362
IUPAC Name 2-iodopropane
SMILES CC(C)I

Specifications

Melting Point -90°C
Density 1.701
Boiling Point 88°C to 90°C
Flash Point 42°C (107°F)
Assay Percent Range 98%, stabilized with copper
Odor Recognizable
Refractive Index 1.4982
Quantity 500 g
UN Number UN2392
Beilstein 1098244
Sensitivity Light sensitive
Merck Index 14,5214
Solubility Information Soluble in water(1.4g/L).
Formula Weight 169.99
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-Iodopropane, stabilized with copper
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241879 Certificate TypeCertificate of Analysis Date08/07/2022 Catalog NumberA15066.14,A15066.36,A15066.22
Lot Number10240936 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog NumberA15066.14,A15066.36,A15066.22
Lot NumberF13Z043 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15066.14,A15066.36,A15066.22
Lot Number10176598 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15066.14,A15066.36,A15066.22
Lot NumberB13Z001 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15066.14,A15066.36,A15066.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.