Learn More
n-Butyllithium, 1.6M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals
Shop All Thermo Scientific ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 14.75, 84.75 |
CAS Max % | 15.25, 85.25 |
Melting Point | -95.0°C |
Color | Pink to White |
Density | 0.6800g/mL |
Boiling Point | 60.0°C to 80.0°C |
Flash Point | −18°C |
Packaging | AcroSeal™ Glass bottle |
Linear Formula | CH3(CH2)3Li |
Quantity | 100 mL |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464899 | Certificate of Analysis | 02/08/2024 | 181271000,181275000,181278000,181270100 |
A0450025 | Certificate of Analysis | 29/04/2024 | 181271000,181275000,181278000,181270100 |
A0451077 | Certificate of Analysis | 29/04/2024 | 181271000,181275000,181278000 |
A0458699 | Certificate of Analysis | 12/12/2023 | 181271000,181275000,181278000,181270100 |
A0435185 | Certificate of Analysis | 31/01/2023 | 181271000,181275000,181278000,181270100 |
Safety and Handling




- n-Butyllithium, 1.6M solution in hexanes
- Danger
- Flammable liquid Category 2
- Substance which\, in contact with water\, emits flammable gases Category 1
- Pyrophoric solid Category 1
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- H250-Catches fire spontaneously if exposed to air.
- H314-Causes severe skin burns and eye damage.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H361f-Suspected of damaging fertility.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contains : Hexane, Butyl lithium
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.