Learn More
2-Amino-4-bromobenzoic acid, 97%, Thermo Scientific Chemicals
30.20€ - 332.66€
Chemical Identifiers
CAS | 20776-50-5 |
---|---|
Molecular Formula | C7H6BrNO2 |
Molecular Weight (g/mol) | 216.034 |
MDL Number | MFCD03618454 |
InChI Key | BNNICQAVXPXQAH-UHFFFAOYSA-N |
Synonym | 4-bromoanthranilic acid, benzoic acid, 2-amino-4-bromo, 5-bromo-2-carboxyaniline, 2-amino-4-bromo-benzoic acid, 2-amino-4-brombenzoic acid, zlchem 336, pubchem13462, acmc-1cb3t, ksc207e6b, pharmabridge p-1344 |
PubChem CID | 88691 |
IUPAC Name | 2-amino-4-bromobenzoic acid |
SMILES | C1=CC(=C(C=C1Br)N)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15482188
|
Thermo Scientific Alfa Aesar
H55829.MD |
250 mg | N/A | ||||||
15462188
|
Thermo Scientific Alfa Aesar
H55829.03 |
1 g |
78.49€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15472188
|
Thermo Scientific Alfa Aesar
H55829.06 |
5 g |
332.66€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (slightly).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
20776-50-5 | |
216.034 | |
BNNICQAVXPXQAH-UHFFFAOYSA-N | |
88691 | |
C1=CC(=C(C=C1Br)N)C(=O)O |
C7H6BrNO2 | |
MFCD03618454 | |
4-bromoanthranilic acid, benzoic acid, 2-amino-4-bromo, 5-bromo-2-carboxyaniline, 2-amino-4-bromo-benzoic acid, 2-amino-4-brombenzoic acid, zlchem 336, pubchem13462, acmc-1cb3t, ksc207e6b, pharmabridge p-1344 | |
2-amino-4-bromobenzoic acid |
Specifications
20776-50-5 | |
C7H6BrNO2 | |
4-bromoanthranilic acid, benzoic acid, 2-amino-4-bromo, 5-bromo-2-carboxyaniline, 2-amino-4-bromo-benzoic acid, 2-amino-4-brombenzoic acid, zlchem 336, pubchem13462, acmc-1cb3t, ksc207e6b, pharmabridge p-1344 | |
BNNICQAVXPXQAH-UHFFFAOYSA-N | |
2-amino-4-bromobenzoic acid | |
88691 | |
97% |
230°C to 234°C | |
MFCD03618454 | |
Soluble in water (slightly). | |
C1=CC(=C(C=C1Br)N)C(=O)O | |
216.034 | |
216.03 | |
2-Amino-4-bromobenzoic acid |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 244-025-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.