missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-bromobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 15482188
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (slightly).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20776-50-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD03618454
InChI Key BNNICQAVXPXQAH-UHFFFAOYSA-N
Synonym 4-bromoanthranilic acid, benzoic acid, 2-amino-4-bromo, 5-bromo-2-carboxyaniline, 2-amino-4-bromo-benzoic acid, 2-amino-4-brombenzoic acid, zlchem 336, pubchem13462, acmc-1cb3t, ksc207e6b, pharmabridge p-1344
PubChem CID 88691
IUPAC Name 2-amino-4-bromobenzoic acid
SMILES C1=CC(=C(C=C1Br)N)C(=O)O

Specifications

Melting Point 230°C to 234°C
Quantity 250 mg
Solubility Information Soluble in water (slightly).
Formula Weight 216.03
Percent Purity 97%
Chemical Name or Material 2-Amino-4-bromobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'