missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ 18/10 Stainless Steel Propeller Stirrers with 4 Fix Blades
Brand: Bochem™ 9005
This item is not returnable.
View return policy
Specifications
50 mm | |
Ideal for homogenizing and suspending | |
400 mm | |
Propeller Stirrer |
8 mm | |
4 Fix blades | |
1 Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bochem™ 18/10 Stainless Steel Propeller Stirrers with 4 Fix Blades > Length: 400mm
Spot an opportunity for improvement?Share a Content Correction