CAS RN 55304-90-0
CAS RN 55304-90-0
IUPAC Name:
(2,6-dichloropyridin-3-yl)methanol
Synonyms:
(2,6-dichloropyridin-3-yl)methanol
Molecular Weight (g/mol):
178.01
Molecular Formula:
C6H5Cl2NO
InChi Key:
FWEVVZQDRPWAND-UHFFFAOYSA-N
SMILES:
OCC1=C(Cl)N=C(Cl)C=C1
1
–
1
of
1
results
1
2,6-Dichloro-3-pyridinemethanol, 95%, Thermo Scientific™
CAS: 55304-90-0 Molecular Formula: C6H5Cl2NO Molecular Weight (g/mol): 178.012 MDL Number: MFCD11036367 InChI Key: FWEVVZQDRPWAND-UHFFFAOYSA-N Synonym: 2,6-dichloropyridin-3-yl methanol,2,6-dichloro-3-pyridinyl methanol,2,6-dichloro-3-hydroxymethyl pyridine,3-pyridinemethanol, 2,6-dichloro,2,6-dichloro-3-hydroxymethylpyridine,2,6-dichloro-3-pyridinemethanol,acmc-209lma,2,6-dichloropyridin-3-yl-methanol,2,6-dichloro-3-pyridyl methan-1-ol,2,6-dichloro-pyridin-3-yl-methanol PubChem CID: 12259932 IUPAC Name: (2,6-dichloropyridin-3-yl)methanol SMILES: C1=CC(=NC(=C1CO)Cl)Cl