Learn More
2,6-Dichloro-3-pyridinemethanol, 95%, Thermo Scientific™
Product Code. 15476867
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55304-90-0
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.012
MDL Number MFCD11036367
InChI Key FWEVVZQDRPWAND-UHFFFAOYSA-N
Synonym 2,6-dichloropyridin-3-yl methanol, 2,6-dichloro-3-pyridinyl methanol, 2,6-dichloro-3-hydroxymethyl pyridine, 3-pyridinemethanol, 2,6-dichloro, 2,6-dichloro-3-hydroxymethylpyridine, 2,6-dichloro-3-pyridinemethanol, acmc-209lma, 2,6-dichloropyridin-3-yl-methanol, 2,6-dichloro-3-pyridyl methan-1-ol, 2,6-dichloro-pyridin-3-yl-methanol
PubChem CID 12259932
IUPAC Name (2,6-dichloropyridin-3-yl)methanol
SMILES C1=CC(=NC(=C1CO)Cl)Cl
Specifications
Melting Point 67°C to 69°C
Quantity 1 g
Formula Weight 178
Percent Purity 95%
Chemical Name or Material 2,6-Dichloro-3-pyridinemethanol