accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 105743-53-1

CAS RN 105743-53-1

IUPAC Name: 2-amino-2,4-dimethylpentanoic acid
Synonyms: 2-methylleucine
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: ARSWQPLPYROOBG-UHFFFAOYNA-N
SMILES: CC(C)CC(C)(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.