missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-alpha-Methylleucine, 98%, 98% ee, Thermo Scientific™
Click to view available options
Quantity:
100 mg
Specifications
Specifications
CAS | 105743-53-1 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | >237°C |
Color | White to Yellow |
Enantiomeric Excess | 98.0% min. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5 to 102.5% |
Molecular Formula | C7H15NO2 |
MDL Number | MFCD01716279 |
Show More |
Safety and Handling
Product Identifier
- (S)-alpha-Methylleucine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction