accessibility menu, dialog, popup

Test

CAS RN 10386-84-2

FFFFBrFFBrFF

CAS RN 10386-84-2

IUPAC Name: 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl
Synonyms: 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl
Molecular Weight (g/mol): 455.93
Molecular Formula: C12Br2F8
InChi Key: YXLMNFVUNLCJJY-UHFFFAOYSA-N
SMILES: FC1=C(F)C(=C(F)C(F)=C1Br)C1=C(F)C(F)=C(Br)C(F)=C1F
Quantity 
  • (1)
  • (6)
  • (1)
Type 
  • (3)
For Use With (Equipment) 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (3)
  • (1)