missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dibromooctafluorobiphenyl in Methyl-tert.butylether 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
Specifications
Specifications
Concentration | 100μg/mL |
CAS | 10386-84-2 |
Packaging | Ampoule |
Quantity | 1 mL |
Type | Organic Single-component Standard Solution |
Molecular Formula | C12Br2F8 |
MDL Number | MFCD00000310 |
InChI Key | YXLMNFVUNLCJJY-UHFFFAOYSA-N |
SMILES | FC1=C(F)C(=C(F)C(F)=C1Br)C1=C(F)C(F)=C(Br)C(F)=C1F |
IUPAC Name | 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'