missing translation for 'onlineSavingsMsg'
Learn More

Internal Standard, 4,4'-Dibromooctafluorobiphenyl, SPEX CertiPrep™

Product Code. 12949164 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
4,4'-Dibromooctafluorobiphenyl10386-84-21000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 10386-84-2, 1634-04-4
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C12Br2F8
MDL Number MFCD00000310
InChI Key YXLMNFVUNLCJJY-UHFFFAOYSA-N
SMILES FC1=C(F)C(=C(F)C(F)=C1Br)C1=C(F)C(F)=C(Br)C(F)=C1F
IUPAC Name 1-bromo-4-(4-bromo-2,3,5,6-tetrafluorophenyl)-2,3,5,6-tetrafluorobenzene
Molecular Weight (g/mol) 455.93
PubChem CID 82600
Methods For US EPA Methods 625, 8150A, 8150B, 8151
Solvent or Matrix MTBE
Chemical Name or Material 4,4'-Dibromooctafluorobiphenyl, SW-846 Methods 8150A, 8150B, 8151
Show More Show Less

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.