missing translation for 'onlineSavingsMsg'
Learn More
Zinc Sulfate Solution, 0.1M, Chem-Lab Product Code.: 11943283

Zinc Sulfate Solution, 0.1M, Chem-Lab

Product Code. 11943283
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 11943283

Brand: Chem Lab CL05.2607.1000

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Used in titrations in laboratory determinations. Zinc Sulfate Solution, 0.1M, Chem-Lab provides a source of zinc for uses compatible with sulfates, including in volumetric solutions that that have been standardized in EDTA.

Zinc Sulfate Heptahydrate is a moderately water and acid soluble Zinc source for uses compatible with sulfates. This solution contains 28.754g per liter (0.1mol/liter).

Specifications

CAS 7446-20-0, 7732-18-5
Color Colorless
CAS Max % 3%
CAS Min % 97%
Physical Form Liquid
Quantity 1 L
Molecular Formula H14O11SZn
Linear Formula H14O11SZn
MDL Number MFCD00149894
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
IUPAC Name zinc;sulfate;heptahydrate
Molecular Weight (g/mol) 287.54
PubChem CID 62640
ChEBI CHEBI:32312
Formula Weight 287.54g/mol
Concentration or Composition (by Analyte or Components) 0.1mol/L (±0.0002 at 20°C)
Odor Odorless
Density 1.01g/mL
Chemical Name or Material Zinc sulfate heptahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc Sulfate Solution, 0.1M, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.