Learn More
Novus Biologicals™ Z-LE(OMe)HD(OMe)-FMK (Caspase 9 Inhibitor)
For use in research applications
Brand:  Novus Biologicals™ NBP2-29398-5MG
 View more versions of this product
Product Code. 18118672
  • 1370.00€ / 5mg
Explore more special offers
Specifications
Tan
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles
Z-LE(OMe)HD(OMe)-FMK (Caspase 9) Inhibitor
Z-LE(OMe)HD(OMe)-FMK (Caspase 9)
In vitro assay
5 mg
690
Crystalline Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Z-LE(OMe)HD(OMe)-FMK (Caspase 9 Inhibitor) > 5mg