missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Z-D(OMe)E(OMe)VD(OMe)-FMK (Caspase 3/7 Inhibitor)

For use in research applications

484.00€ - 1370.00€

Specifications

Color White
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18769823
View Documents
Novus Biologicals™
NBP2-29396
484.00€
1mg
Please to purchase this item. Need a web account? Register with us today!
18136913
View Documents
Novus Biologicals™
NBP2-29396-5MG
5 mg
1370.00€
5mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

White
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Z-D(OMe)E(OMe)VD(OMe)-FMK (Caspase 3/7 Inhibitor)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.