missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation XBridge BEH130 Prep C18 Column
The XBridge BEH130 C18 Peptide Separation Technology (PST) preparative columns are optimized and QC tested for peptide separations.
Brand: Waters Corporation 186003662

This item is not returnable.
View return policy
Description
- The small-pore (130 Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
- The Optimum Bed Denisity (OBD) hardware format ensures superior column lifetime under a wide variety of operating conditions.
Specifications
30 mm | |
6000 psi (415 Bar) | |
10 μm | |
1/Ea. | |
Yes | |
Peptides |
250 mm | |
Spherical | |
130 Å | |
L1 | |
Column |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation XBridge BEH130 Prep C18 Column
Spot an opportunity for improvement?Share a Content Correction