missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WZ 4003

Potent and selective NUAK1/2 inhibitor

260.00€ - 1015.00€

Specifications

Purity >98%
Product Type WZ 4003
Molecular Weight (g/mol) 496.99
Inhibitors WZ 4003
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18750278
View Documents
Tocris Bioscience™
5177/50
50 mg
1015.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
18729937
View Documents
Tocris Bioscience™
5177/10
10 mg
260.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent and selective NUAK1/2 inhibitor (IC50 values are 20 and 100 nM respectively). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells.
Specifications

Specifications

>98%
WZ 4003
496.99
WZ 4003
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.