Learn More
Tocris Bioscience™ WZ 4003
Potent and selective NUAK1/2 inhibitor
Brand: Tocris Bioscience™ 5177/50
This item is not returnable.
View return policy
Description
WZ 4003 is a potent and selective NUAK1/2 inhibitor (IC50 values are 20 and 100 nM respectively). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells.Specifications
>98% | |
WZ 4003 | |
WZ 4003 |
50 mg | |
496.99 |