Learn More
Tocris Bioscience™ WZ 4003
Potent and selective NUAK1/2 inhibitor
Brand:  Tocris Bioscience™ 5177/50
 View more versions of this product
Product Code. 18750278
  • 1015.00€ / 50mg
Explore more special offers
Description
Description
WZ 4003 is a potent and selective NUAK1/2 inhibitor (IC50 values are 20 and 100 nM respectively). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells.
Specifications
Specifications
>98%
WZ 4003
WZ 4003
50 mg
496.99