missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper Product Code.: 12349719
Add to basket

DWK Life Sciences Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper

Product Code. 12349719 Shop All Wheaton Science Products Products
100 mL
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 12349719

Brand: DWK Life Sciences W215255

Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for shelf storage

  • Amber borosilicate glass bottle with narrow mouth
  • Protects light-sensitive reagents and solutions
  • Ground-glass octagon flat stopper provides excellent seal; protects top lip of bottle from dust

TRUSTED_SUSTAINABILITY

Specifications

Capacity 3.38 oz., 100mL
Capacity (Metric) 100 mL
Capacity (English) 3.38 oz.
Color Amber
Diameter (English) 2.08 in.
Material Glass
Closure Color Amber
Mouth Narrow
Height (Metric) 115 mm
Height (English) 4.52 in.
Closure Material Borosilicate Glass
Diameter (Metric) 53 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.